Learn More
Aviva Systems Biology Corp PAWR Antibody - middle region (ARP32735_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32735_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PAWR. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PAWR. Alternate names: PAR4, Par-4ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52869