Learn More
Aviva Systems Biology Corp PAX2 Antibody - middle region (P100859_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100859_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PAX2. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62748