Learn More
Aviva Systems Biology Corp PAX4 Antibody - middle region (ARP32064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PAX4. Alternate names: KPD, MGC129960, MODY9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54439