Learn More
Aviva Systems Biology Corp PAX4 Antibody - N-terminal region (ARP32737_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32737_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAX4. Alternate names: MGC129960, KPD, MODY9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54734