Learn More
Aviva Systems Biology Corp PAX5 Antibody - N-terminal region (ARP34686_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34686_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PAX5. Alternate names: BSAPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55513