Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5080MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM612. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Pancreas, Cerebellum or Stomach. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment (N-terminus; aa 1-300) of human PAX6 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 47kDa.Chromosome: 11p13. Gene Symbol: PAX6. Gene ID: 5080. Ensemble: ENSG00000007372. Uniprot: NP_000271.
Catalog No. 50-188-5031