Learn More
Aviva Systems Biology Corp PAX6 Antibody - N-terminal region (ARP38062_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38062_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAX6. Alternate names: AN, AN2, MGDA, WAGR, D11S812EApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56506