Learn More
Aviva Systems Biology Corp PAX6 Antibody - N-terminal region (ARP32741_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32741_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAX6. Alternate names: AN, AN2, D11S812E, MGC17209, MGDA, WAGRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54735