Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5081MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: PAX7/497. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HeLa, Jurkat, A431, MCF-7, U2OS or NIH/3T3 cells. Rhabdomyosarcoma (RMS). Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment (aa301-505) of human PAX7 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 57kDa.Chromosome: 1p36.13. Gene Symbol: PAX7. Gene ID: 5081. Ensemble: ENSG00000009709. Uniprot: NP_039236.
Catalog No. 50-188-5032