Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5081MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: SPM613. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Rhabdomyosarcoma (RMS) or Testicular Carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment (aa300-600) of human PAX7 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 57kDa.Chromosome: 1p36.13. Gene Symbol: PAX7. Gene ID: 5081. Ensemble: ENSG00000009709. Uniprot: NP_039236.
Catalog No. 50-188-5034