Learn More
Aviva Systems Biology Corp PAX7 Antibody - C-terminal region (ARP32742_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32742_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of PAX7. Alternate names: FLJ37460, HUP1, PAX7B, RMS2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54736