Learn More
Biocare Medical, LLC PAX8 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CP379CK
Encompass_Preferred
PAX8 Rabbit Antibody is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically encode proteins which contain a paired box domain, an octapeptide, and a paired-type homeodomain. This family plays critical roles during fetal development and cancer growth. PAX8 is involved in kidney cell differentiation, thyroid development, or thyroid dysgenesis. Studies show that expression of the PAX8 gene was found in 89% of analyzed tumor samples. The expression of the PAX8 target genes were found in all normal renal samples. PAX8 has been shown to be expressed in three of the most common types of renal cell carcinoma including clear cell, chromophobe and papillary carcinoma but negative for uroepithelial carcinoma of renal pelvis. PAX8 stains nuclei exclusively and performs well in formalin-fixed paraffinembedded tissues. PAX8 has been shown to be a superior marker compared to the Renal Cell Carcinoma Marker (RCC).
Catalog No. 50-827-83