Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7849MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: PAX8/1491. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: Raji cells. Renal Cell Carcinoma (RCC). Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human PAX8 fragment (aa60-261). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 62kDa.Chromosome: 2q13. Gene Symbol: PAX8. Gene ID: 7849. Ensemble: ENSG00000125618. Uniprot: XP_016860367.
Catalog No. 50-188-5300