Learn More
Aviva Systems Biology Corp PAX8 Antibody - N-terminal region (ARP48088_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48088_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAX8. Alternate names: -ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59716