Learn More
Cayman Chemical Pax8CTerm RabbIt Monclon 100uL
SDP
Catalog No. 502913474
Encompass_Preferred
A flavone with diverse biological activities binds to rat adenosine A1 rat A2A and human A3 receptors (Kis 1 2 3 and 1 72 UM respectively in radioligand binding assays) inhibits DPP-4 (IC50 0 43 UM) active against the NF54 strain of P falciparum (IC50 16 9 UM) inhibits proliferation of AGS HT-29 Saos-2 and WEHI 164 cells (IC50s 14 4 13 1 38 5 and 40 7 UM respectively increases the number of entries into and percentage of time spent in the open arms of the elevated plus maze in mice at 10 mg/kg
Catalog No. 50-291-3474 Supplier Cayman Chemical Supplier No. 35615100 UL
A flavone with diverse biological activities binds to rat adenosine A1 rat A2A and human A3 receptors (Kis 1 2 3 and 1 72 UM respectively in radioligand binding assays) inhibits DPP-4 (IC50 0 43 UM) active against the NF54 strain of P falciparum (IC50 16 9 UM) inhibits proliferation of AGS HT-29 Saos-2 and WEHI 164 cells (IC50s 14 4 13 1 38 5 and 40 7 UM respectively increases the number of entries into and percentage of time spent in the open arms of the elevated plus maze in mice at 10 mg/kg