Learn More
Aviva Systems Biology Corp PAX9 Antibody - N-terminal region (ARP34270_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34270_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAX9. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PAX9. Alternate names: STHAG3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55319