Learn More
Andwin Scientific PAXGENE BLD RNA TUBES CS/100
SDP
Catalog No. NC9413258
Encompass_Preferred
The PAXgene Blood RNA Plastic Tube features a clear Hemoguard closure. It has a volume capacity of 2.5 mL. This product also has a paper label that makes identification and organization easier. This is sold in a case of 100.
This item is not returnable. View return policy
Catalog No. NC9413258 Supplier Andwin Scientific Supplier No. 77776026BD
The PAXgene Blood RNA Plastic Tube features a clear Hemoguard closure. It has a volume capacity of 2.5 mL. This product also has a paper label that makes identification and organization easier. This is sold in a case of 100.