Learn More
Cayman Chemical PaxIlInaN-Term RabbIt 100uL
SDP
Catalog No. 502950590
Encompass_Preferred
A partial agonist of muscarinic acetylcholine receptors (Kis 5 1 5 0.2 and 7.6 UM for M1-5 receptors respectively in radioligand binding assays) induces contraction of isolated human umbilical veins (EC50 1.23 UM) intrathecal administration inhibits GNTI-induced compulsive hindlimb neck-scratching behavior in mice enhances spontaneous working memory in the Y maze in wild-type mice and reverses learning and memory impairments induced by bulbectomy in mice in a passive avoidance test
Catalog No. 50-295-0590 Supplier Cayman Chemical Supplier No. 32209100 UL
A partial agonist of muscarinic acetylcholine receptors (Kis 5 1 5 0.2 and 7.6 UM for M1-5 receptors respectively in radioligand binding assays) induces contraction of isolated human umbilical veins (EC50 1.23 UM) intrathecal administration inhibits GNTI-induced compulsive hindlimb neck-scratching behavior in mice enhances spontaneous working memory in the Y maze in wild-type mice and reverses learning and memory impairments induced by bulbectomy in mice in a passive avoidance test