Learn More
ECM Bioscience Paxillin Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience PK6070
Encompass_Preferred
The paxillin phospho-regulation antibody sampler kit can be used to examine phosphorylation of paxillin at Tyr-31, Ser-83, and Ser-178. The kit also includes monoclonal and polyclonal antibodies to monitor total paxillin expression levels. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8773