Learn More
Aviva Systems Biology Corp PAXIP1 Antibody - N-terminal region (ARP51642_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51642_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAXIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAXIP1. Alternate names: PTIP, TNRC2, CAGF28, CAGF29, PACIP1, PAXIP1L, FLJ41049ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60575