Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pazopanib-d6 | 1219592-01-4 | MFCD11110377 | 98.0% | 443.55 g/mol | C21H17D6N7O2S | 1 MG
SDP

Catalog No. 5000248527
Encompass_Preferred

Pazopanib-d6 is the deuterium-labeled analogue of pazopanib supplied as a research reagent for use as an internal standard in analytical and pharmacokinetic studies. The material is provided as a small weighed quantity of dry powder and should be stored under recommended cold conditions.

  • Deuterium-labeled internal standard suitable for LC-MS/MS and PK assays
  • High purity (98.0%) for reliable quantitative analysis
  • Chemical formula C21H17D6N7O2S and molecular weight 443.55 g/mol
  • Available in small weighed quantities for analytical workflows
  • Store powder at -20°C for long-term stability

Catalog No. 50-002-48527 Supplier Medchemexpress LLC Supplier No. HY10208S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pazopanib-d6 is the deuterium-labeled analogue of pazopanib supplied as a research reagent for use as an internal standard in analytical and pharmacokinetic studies. The material is provided as a small weighed quantity of dry powder and should be stored under recommended cold conditions.

  • Deuterium-labeled internal standard suitable for LC-MS/MS and PK assays
  • High purity (98.0%) for reliable quantitative analysis
  • Chemical formula C21H17D6N7O2S and molecular weight 443.55 g/mol
  • Available in small weighed quantities for analytical workflows
  • Store powder at -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.