Learn More
Selleck Chemical LLC Pazopanib HCl S1035-10mg
SDP
Catalog No. 503108144
Encompass_Preferred
Pazopanib HCl is a novel multi-target inhibitor of VEGFR1 VEGFR2 VEGFR3 PDGFR FGFR c-Kit and c-Fms with IC50 of 10 nM 30 nM 47 nM 84 nM 74 nM 140 nM and 146 nM in cell-free assays respectively Pazopanib induces autophagic Type II cell death
Catalog No. 50-310-8144 Supplier Selleck Chemical LLC Supplier No. S103510MG
Pazopanib HCl is a novel multi-target inhibitor of VEGFR1 VEGFR2 VEGFR3 PDGFR FGFR c-Kit and c-Fms with IC50 of 10 nM 30 nM 47 nM 84 nM 74 nM 140 nM and 146 nM in cell-free assays respectively Pazopanib induces autophagic Type II cell death