Learn More
Selleck Chemical LLC Pazopanib HCl (GW786034 HCl) 100mg 635702-64-6
SDP
Catalog No. 506213
Encompass_Preferred
Pazopanib HCl (GW786034 HCl) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFR, FGFR, c-Kit and c-Fms with IC50 of 10 nM, 30 nM, 47 nM, 84 nM, 74 nM, 140 nM and 146 nM in cell-free assays, respectively. Pazopanib induces autophagic Type II cell death. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-621-3 Supplier Selleck Chemical LLC Supplier No. S1035100MG
Pazopanib HCl (GW786034 HCl) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFR, FGFR, c-Kit and c-Fms with IC50 of 10 nM, 30 nM, 47 nM, 84 nM, 74 nM, 140 nM and 146 nM in cell-free assays, respectively. Pazopanib induces autophagic Type II cell death. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.