Learn More
Medchemexpress LLC Pazopanib hydrochloride | 635702-64-6 | 100.0% | 1 ML
SDP
Catalog No. 5000232171
Encompass_Preferred
Pazopanib hydrochloride is a high-purity research reagent that functions as a multi-target tyrosine kinase inhibitor. It is provided in both solid quantities and as a ready-to-use 10 mM solution in DMSO (1 mL) for convenient use in biochemical and cellular assays.
  • Multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRβ, c-Kit, FGFR1, and c-Fms.
  • High purity for analytical and research applications (99.99%).
  • Available as solid packs and as a 10 mM solution in DMSO, 1 mL.
  • Ready-to-use solution simplifies dosing and assay setup.
  • Suitable for biochemical, cellular, and analytical studies.
Catalog No. 50-002-32171 Supplier Medchemexpress LLC Supplier No. HY1200910MM 1ML
Pazopanib hydrochloride is a high-purity research reagent that functions as a multi-target tyrosine kinase inhibitor. It is provided in both solid quantities and as a ready-to-use 10 mM solution in DMSO (1 mL) for convenient use in biochemical and cellular assays.
  • Multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRβ, c-Kit, FGFR1, and c-Fms.
  • High purity for analytical and research applications (99.99%).
  • Available as solid packs and as a 10 mM solution in DMSO, 1 mL.
  • Ready-to-use solution simplifies dosing and assay setup.
  • Suitable for biochemical, cellular, and analytical studies.