Learn More
Medchemexpress LLC Pazopanib hydrochloride | 635702-64-6 | 100.0% | 50 MG
SDP
Catalog No. 5000232172
Encompass_Preferred
Pazopanib hydrochloride is the hydrochloride salt of pazopanib, a small-molecule multi-target tyrosine kinase inhibitor active against VEGFR1/2/3, PDGFRβ, c-Kit, and FGFR1. It is supplied as a research reagent for in vitro pharmacology, cell-based assays, and preclinical studies.
  • High purity suitable for research applications.
  • Available as solid and DMSO solution formats for flexible use.
  • Useful for kinase inhibition, cell signaling, and oncology research.
  • Offered in multiple pack sizes to support screening and preclinical work.
Catalog No. 50-002-32172 Supplier Medchemexpress LLC Supplier No. HY1200950MG
Pazopanib hydrochloride is the hydrochloride salt of pazopanib, a small-molecule multi-target tyrosine kinase inhibitor active against VEGFR1/2/3, PDGFRβ, c-Kit, and FGFR1. It is supplied as a research reagent for in vitro pharmacology, cell-based assays, and preclinical studies.
  • High purity suitable for research applications.
  • Available as solid and DMSO solution formats for flexible use.
  • Useful for kinase inhibition, cell signaling, and oncology research.
  • Offered in multiple pack sizes to support screening and preclinical work.