Learn More
Biolegend Pacific Blue™ anti-human CD11c
Supplier:  Biolegend 301626
Encompass_Preferred
Pacific Blue™ anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. 50-165-732