Learn More
Biolegend Pacific Blue™ anti-human CD15 (SSEA-1)
Supplier:  Biolegend 323022
Encompass_Preferred
Pacific Blue™ anti-human CD15 (SSEA-1) [W6D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-162