Learn More
Biolegend Pacific Blue™ anti-human CD2
Supplier:  Biolegend 309216
Encompass_Preferred
Pacific Blue™ anti-human CD2 [TS1/8]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-166-501