Learn More
Biolegend Pacific Blue™ anti-human CD3
Supplier:  Biolegend 317313
Encompass_Preferred
Pacific Blue™ anti-human CD3 [OKT3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 ug
Catalog No. 50-166-911