Learn More
Biolegend Pacific Blue™ anti-human CD4
Supplier:  Biolegend 317423
Encompass_Preferred
Pacific Blue™ anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 25 ug
Catalog No. 50-166-934