Learn More
Biolegend Pacific Blue™ anti-human CD45
Supplier:  Biolegend 304021
Encompass_Preferred
Pacific Blue™ anti-human CD45 [HI30]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 ug
Catalog No. 50-166-062