Learn More
Biolegend Pacific Blue™ anti-human CD89
Supplier:  Biolegend 354119
Encompass_Preferred
Pacific Blue™ anti-human CD89 [A59]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-207-7321