Learn More
Biolegend Pacific Blue™ anti-human FOXP3
Supplier:  Biolegend 320215
Encompass_Preferred
Pacific Blue™ anti-human FOXP3 [259D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee; Apps: ICFC; Size: 25 tests
Catalog No. 50-167-020