Learn More
Biolegend Pacific Blue™ anti-human Granzyme A
Supplier:  Biolegend 507207
Encompass_Preferred
Pacific Blue™ anti-human Granzyme A [CB9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 ug
Catalog No. 50-170-018