Learn More
Biolegend Pacific Blue™ anti-human HLA-A2
Supplier:  Biolegend 343312
Encompass_Preferred
Pacific Blue™ anti-human HLA-A2 [BB7.2]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-403-363