Learn More
Biolegend Pacific Blue™ anti-human IFN-γ
Supplier:  Biolegend 506526
Encompass_Preferred
Pacific Blue™ anti-human IFN-γ [B27]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, African Green Monkey, Pigtailed Macaque, Sooty Mangabey; Apps: ICFC; Size: 100 tests
Catalog No. 50-208-5758