Learn More
Biolegend Pacific Blue™ anti-human IL-1β
Supplier:  Biolegend 511709
Encompass_Preferred
Pacific Blue™ anti-human IL-1β [H1b-98]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 ug
Catalog No. 50-170-105