Learn More
Biolegend Pacific Blue™ anti-human/mouse Granzyme B [GB11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Supplier:  Biolegend 515407
Encompass_Preferred
Pacific Blue™ anti-human/mouse Granzyme B [GB11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-237-6730