Learn More
Biolegend Pacific Blue™ anti-human/mouse/rat CD278 (ICOS)
Supplier:  Biolegend 313522
Encompass_Preferred
Pacific Blue™ anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Mouse, Rat, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 ug
Catalog No. 50-166-755