Learn More
Biolegend Pacific Blue™ anti-human Perforin
Supplier:  Biolegend 308118
Encompass_Preferred
Pacific Blue™ anti-human Perforin [dG9]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Bovine (Cow, Cattle); Apps: ICFC; Size: 100 tests
Catalog No. 50-166-465