Learn More
Biolegend Pacific Blue™ anti-human CD45
Supplier:  Biolegend 368539
Encompass_Preferred
Pacific Blue™ anti-human CD45 [2D1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1433697