Learn More
Biolegend Pacific Blue™ anti-IRF4
Supplier:  Biolegend 646418
Encompass_Preferred
Pacific Blue™ anti-IRF4 [IRF4.3E4]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human; Apps: ICFC; Size: 100 ug
Catalog No. 50-208-6944