Learn More
Biolegend Pacific Blue™ anti-mouse CD117 (c-kit)
Supplier:  Biolegend 105820
Encompass_Preferred
Pacific Blue™ anti-mouse CD117 (c-kit) [2B8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-604-861