Learn More
Biolegend Pacific Blue™ anti-mouse CD19
Supplier:  Biolegend 115526
Encompass_Preferred
Pacific Blue™ anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-553