Learn More
Biolegend Pacific Blue™ anti-mouse CD25
Supplier:  Biolegend 102021
Encompass_Preferred
Pacific Blue™ anti-mouse CD25 [PC61]; Isotype: Rat IgG1, λ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-628