Learn More
Biolegend Pacific Blue™ anti-mouse CD31
Supplier:  Biolegend 102421
Encompass_Preferred
Pacific Blue™ anti-mouse CD31 [390]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-692