Learn More
Biolegend Pacific Blue™ anti-mouse CD38
Supplier:  Biolegend 102719
Encompass_Preferred
Pacific Blue™ anti-mouse CD38 [90]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-730