Learn More
Biolegend Pacific Blue™ anti-mouse CD4
Supplier:  Biolegend 100427
Encompass_Preferred
Pacific Blue™ anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC0127288