Learn More
Biolegend Pacific Blue™ anti-mouse CD54
Supplier:  Biolegend 116115
Encompass_Preferred
Pacific Blue™ anti-mouse CD54 [YN1/1.7.4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-614