Learn More
Biolegend Pacific Blue™ anti-mouse IFN-γ
Supplier:  Biolegend 505818
Encompass_Preferred
Pacific Blue™ anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-169-929